
4-tert-Octylphenol diethoxylate solution

Synonym(s):
2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethoxy}ethanol
Empirical Formula (Hill Notation):
C18H30O3
CAS Number:
Molecular Weight:
294.43
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing