
1H-Imidazole-4-sulfonyl chloride

Empirical Formula (Hill Notation):
C3H3ClN2O2S
Molecular Weight:
166.59
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing