Merck
  • Significance of the CAG repeat length in the androgen receptor gene (AR) for the transactivation function of an M780I mutant AR.
Significance of the CAG repeat length in the androgen receptor gene (AR) for the transactivation function of an M780I mutant AR.
Human genetics (1999-05-14)
I Knoke, A Allera, P Wieacker
RESUMEN
Mutations in the androgen receptor gene (AR) cause a wide spectrum of androgen insensitivity syndromes (AIS). Mutation analysis of patients with AIS has revealed that the same missense mutation of the AR gene can give rise to strongly divergent phenotypes suggesting the influence of modifying factors. The polymorphic CAG repeat in the first exon of the AR gene may be such a modifying factor. The influence of the length of the CAG repeat on the transactivation function of the M780I-mutant AR (causing partial and complete AIS) has been determined by cotransfection of HeLa cells with various CAG-AR expression vectors and a highly androgen-responsive luciferase reporter gene construct. The transcriptional activity of the M780I mutant AR can be, in contrast to the wild-type AR, considerably enhanced by non-physiologically high androgen concentrations. Furthermore, an inverse relationship between the number of the CAG repeats in the mutant AR and its activity has been observed.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Roche
DOTAP Liposomal Transfection Reagent, >99% (TLC), liquid, suitable for transfection