Merck
The mechanism of antiparallel β-sheet formation based on conditioned self-avoiding walk.
The European physical journal. E, Soft matter (2012-04-25)
Boon Chong Goh, Hon Wai Leong, Xiaohui Qu, Lock Yue Chew
RESUMEN
By introducing an additional hydrogen bond to hydrogen bond interaction in the force field of the CSAW (Conditioned Self-Avoiding Walk) model, we investigate into the mechanism of antiparallel β-sheet formation based on the folding of a short polyalanine in gas phase. Through our numerical simulation, we detect the possible presence of a transient helix during β-sheet formation, whose presence is shown to have slowed the formation of β-sheets by an order of magnitude. While we observe the mechanisms of nucleation, zipping and induction that drives the formation of a β-sheet, we uncover a new mechanism that involves transient β-turns and short β-sheets during the formation of long β-sheets. Our results have enabled us to provide an overview on the mechanisms of β-sheet formation via two main folding pathways: slow folding through the intermediate state of transient helix, and fast folding from the nucleation of β-turn.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Poly-DL-alanine, mol wt 1,000-5,000