Merck
Photodegradation of Orange II by mesoporous TiO2.
Journal of environmental monitoring : JEM (2011-08-13)
Liyuan Kuang, Yaping Zhao, Lu Liu
RESUMEN
Mesoporous TiO(2) microspheres were prepared by a hydrothermal reaction and are characterized in this paper. Decoloration and mineralization during photodegradation of Orange II by mesoporous TiO(2) at different pH values, formation of sulfate, relative luminosity to luminous bacteria and recycling experiments of the catalyst were studied. The FTIR results further suggested that the novel mesoporous TiO(2) can not only decolor and mineralize dyes completely but also can be effectively reused several times. On the basis of the research, mesoporous TiO(2) would be a promising photocatalyst for practical use.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Orange II sodium salt, (Certified by the Biological Stain Commission), Dye content ≥85 %
Supelco
Orange II sodium salt, analytical standard