Merck
The glucocorticoid-Angptl4-ceramide axis induces insulin resistance through PP2A and PKCζ.
Science signaling (2017-07-27)
Tzu-Chieh Chen, Daniel I Benjamin, Taiyi Kuo, Rebecca A Lee, Mei-Lan Li, Darryl J Mar, Damian E Costello, Daniel K Nomura, Jen-Chywan Wang
RESUMEN
Chronic glucocorticoid exposure is associated with the development of insulin resistance. We showed that glucocorticoid-induced insulin resistance was attenuated upon ablation of Angptl4, a glucocorticoid target gene encoding the secreted protein angiopoietin-like 4, which mediates glucocorticoid-induced lipolysis in white adipose tissue. Through metabolomic profiling, we revealed that glucocorticoid treatment increased hepatic ceramide concentrations by inducing enzymes in the ceramide synthetic pathway in an Angptl4-dependent manner. Angptl4 was also required for glucocorticoids to stimulate the activities of the downstream effectors of ceramide, protein phosphatase 2A (PP2A) and protein kinase Cζ (PKCζ). We further showed that knockdown of PP2A or inhibition of PKCζ or ceramide synthesis prevented glucocorticoid-induced glucose intolerance in wild-type mice. Moreover, the inhibition of PKCζ or ceramide synthesis did not further improve glucose tolerance in Angptl4-/- mice, suggesting that these molecules were major downstream effectors of Angptl4. Overall, our study demonstrates the key role of Angptl4 in glucocorticoid-augmented hepatic ceramide production that induces whole-body insulin resistance.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
trans-(±)-ACPD monohydrate, solid, ≥97% (NMR)