Merck
  • Hydroalcoholic extract from Origanum vulgare induces a combined anti-mycobacterial and anti-inflammatory response in innate immune cells.
Hydroalcoholic extract from Origanum vulgare induces a combined anti-mycobacterial and anti-inflammatory response in innate immune cells.
PloS one (2019-03-05)
Federica De Santis, Noemi Poerio, Angelo Gismondi, Valentina Nanni, Gabriele Di Marco, Roberto Nisini, Maria Cristina Thaller, Antonella Canini, Maurizio Fraziano
RESUMEN
In nature, many plants or their extracted compounds have been found to possess anti-inflammatory features and therapeutic properties against infectious as well as non-infectious diseases, including cancer. In this study, we analysed the immunomodulatory effects on innate immune cells of hydroalcoholic extract from Origanum vulgare L. ssp. hirtum (HyE-Ov), a plant traditionally known for its anti-oxidative properties. The effects of HyE-Ov were tested on human monocyte derived dendritic cells (DC), type-1 (M1) and type-2 macrophages (M2) infected with M. bovis Bacille Calmette-Guérin (BCG), used as a model of persistent intracellular bacterium. DC, M1 and M2 treated with HyE-Ov significantly enhanced their mycobactericidal activity, which was associated with phagosomal acidification in M1 and M2 and increase of phagosomal, but not mitochondrial ROS production in M1, M2, and DC. Treatment of BCG-infected DC with HyE-Ov significantly reduced TNF-α and IL-12 production and increased TGF-β synthesis. Finally, experiments were repeated using eight different HPLC fractions of HyE-Ov. Results showed that the capability to activate anti-microbial and anti-inflammatory response is shared by different fractions, suggesting that diverse bioactive molecules are present within the hydroalcoholic extract. Altogether, these results show that HyE-Ov promotes anti-mycobacterial innate immunity and limits inflammatory response in vitro and suggest that this plant extract may be exploitable as phytocomplex or nutraceutical for novel host-directed therapeutic approaches.