Merck
Monitoring Hydrogenation Reactions using Benchtop 2D NMR with Extraordinary Sensitivity and Spectral Resolution.
ChemistryOpen (2019-03-01)
Dariusz Gołowicz, Krzysztof Kazimierczuk, Mateusz Urbańczyk, Tomasz Ratajczyk
RESUMEN
Low-field benchtop nuclear magnetic resonance (BT-NMR) spectrometers with Halbach magnets are being increasingly used in science and industry as cost-efficient tools for the monitoring of chemical reactions, including hydrogenation. However, their use of low-field magnets limits both resolution and sensitivity. In this paper, we show that it is possible to alleviate these two problems through the combination of parahydrogen-induced polarization (PHIP) and fast correlation spectroscopy with time-resolved non-uniform sampling (TR-NUS). PHIP can enhance NMR signals so that substrates are easily detectable on BT-NMR spectrometers. The interleaved acquisition of one- and two-dimensional spectra with TR-NUS provides unique insight into the consecutive moments of hydrogenation reactions, with a spectral resolution unachievable in a standard approach. We illustrate the potential of the technique with two examples: the hydrogenation of ethylphenyl propiolate and the hydrogenation of a mixture of two substrates - ethylphenyl propiolate and ethyl 2-butynoate.