Merck
Exceptionally strong multiphoton-excited blue photoluminescence and lasing from ladder-type oligo(p-phenylene)s.
Journal of the American Chemical Society (2012-04-24)
Hai Hua Fan, Lei Guo, King Fai Li, Man Shing Wong, Kok Wai Cheah
RESUMEN
We report the synthesis and investigation of multiphoton absorption properties of a novel series of diphenylamino-end-capped ladder-type oligo(p-phenylene)s which exhibit greatly enhanced and efficient multiphoton (from two- to five-photon) upconverted blue photoluminescence with which the record-high intrinsic three-photon absorption cross-section of 4.56 × 10(-76) cm(6) s(2) in the femtosecond regime has been obtained. Exceptionally efficient two- to five-photon-excited lasing in the blue region has also been demonstrated in which the highest two-photon-excited lasing efficiency of 0.34% has been achieved.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-(Diphenylamino)phenylboronic acid, ≥95%