HPLC Analysis of Fenofibric Acid Enantiomers on Astec® CYCLOBOND I 2000
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] 0.1% triethylamine acetate, pH 4.0; [B] acetonitrile; (20:80, A:B)
flow rate
1 mL/min
column temp.
22 °C
detector
UV, 254 nm
injection
1 μL
sample
3 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null