Merck
  • A soluble form of CD80 enhances antitumor immunity by neutralizing programmed death ligand-1 and simultaneously providing costimulation.
A soluble form of CD80 enhances antitumor immunity by neutralizing programmed death ligand-1 and simultaneously providing costimulation.
Cancer immunology research (2014-05-14)
Samuel T Haile, Lucas A Horn, Suzanne Ostrand-Rosenberg
ABSTRACT
Tumor cells use various methods of immunosuppression to overcome antitumor immunity. One such method is that of programmed death ligand-1 (PD-L1 or B7-H1), which upon binding its receptor PD-1 on T cells triggers apoptotic death of the activated T cells. Overexpression of the costimulatory molecule CD80 on PD-L1(+) tumor cells, or inclusion of a soluble form of CD80 (CD80-Fc), maintains the activation of PD-1(+)-activated T cells. Using T cells from CD28-deficient mice and antibodies to block CD28 on human T cells, we now report that a soluble form of CD80 mediates this effect by simultaneously neutralizing PD-1-PD-L1-mediated immunosuppression and by providing CD80-CD28 costimulation, and is more effective than antibodies to PD-L1 or PD-1 in maintaining IFNγ production by PD-1(+) activated T cells. Therefore, soluble CD80 may be a more effective therapeutic than these checkpoint antibodies for facilitating the development and maintenance of antitumor immunity because it has the dual functions of preventing PD-L1-mediated immunosuppression and simultaneously delivering the second signal for T-cell activation.
Sigma-Aldrich
Human IFN γ ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IFNA1 / Interferon Alpha-1/13 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Human IFN γ ELISA Kit, for cell and tissue lysates