Merck
  • The TLR7 agonist imiquimod as an adjuvant for radiotherapy-elicited in situ vaccination against breast cancer.
The TLR7 agonist imiquimod as an adjuvant for radiotherapy-elicited in situ vaccination against breast cancer.
Oncoimmunology (2014-01-10)
Sandra Demaria, Claire Vanpouille-Box, Silvia C Formenti, Sylvia Adams
ABSTRACT
Radiotherapy can convert malignant cells into an in situ anticancer vaccine, but is often inadequate at generating sufficient pro-inflammatory signals to optimally activate innate and adaptive immune responses. Topical imiquimod is a powerful pro-inflammatory agent with clinical activity against superficial skin cancers. These two modalities appear to complement each other, hence achieving local and systemic tumor control.
Sigma-Aldrich
Bropirimine, ≥98% (HPLC)