Merck
  • Influence of cytochrome P-450 type on the pattern of conjugation of 7-hydroxycoumarin generated from 7-alkoxycoumarins.
Influence of cytochrome P-450 type on the pattern of conjugation of 7-hydroxycoumarin generated from 7-alkoxycoumarins.
Xenobiotica; the fate of foreign compounds in biological systems (1984-11-01)
P Paterson, J R Fry, S A Horner
ABSTRACT
Pretreatment of rats with phenobarbitone increased hepatic microsomal 7-methoxy-and 7-ethoxy-coumarin O-dealkylase activities. Pretreatment with beta-naphthoflavone increased only the 7-ethoxycoumarin O-dealkylase activity. The addition of metyrapone in vitro inhibited the O-dealkylations to different extents. Similar results were obtained with diphenyloxazole and ethanol. These results are taken to indicate that different forms of cytochrome P-450 are involved in the O-dealkylation of these two substrates. The pattern of metabolism (Phase I and Phase II) of each alkoxycoumarin in rat isolated hepatocytes was very similar. The sulphate conjugate was the major metabolite produced, the amount of which approached a plateau as the rate of O-dealkylation increased. It is concluded that the type of cytochrome P-450 involved in the initial Phase I metabolism does not influence the subsequent pattern of conjugation.
Sigma-Aldrich
7-Methoxycoumarin, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
7-Methoxycoumarin, ≥98%