Merck
  • Selective phenolic acylation of 10-hydroxycamptothecin using poly (ethylene glycol) carboxylic acid.
Selective phenolic acylation of 10-hydroxycamptothecin using poly (ethylene glycol) carboxylic acid.
Bioorganic & medicinal chemistry letters (2003-02-05)
Richard B Greenwald, Yun H Choe, Dechun Wu
ABSTRACT
Selective acylation of the phenolic hydroxyl group of 10-hydroxycamptothecin has been accomplished using phenyl dichlorophosphate. Additional modification of the 10-OH as an ether permits a 20-acyl derivative to be synthesized. This result along with data from a 6-hydroxyquinoline model strongly suggests that powerful intermolecular hydrogen bonding exists in the parent molecule.
Sigma-Aldrich
6-Hydroxyquinoline, 95%