Merck
  • Polyhydroxyalkanoates production by a mixed photosynthetic consortium of bacteria and algae.
Polyhydroxyalkanoates production by a mixed photosynthetic consortium of bacteria and algae.
Bioresource technology (2013-02-13)
J C Fradinho, J M B Domingos, G Carvalho, A Oehmen, M A M Reis
ABSTRACT
For the first time, a mixed photosynthetic culture (MPC) consisting of a consortium of bacteria and algae was investigated for its capacity to accumulate polyhydroxyalkanoates (PHA). The culture was subjected to a feast and famine regime in an illuminated environment without supplying oxygen or any other electron acceptor. The MPC accumulated PHA during the feast phase and consumed it in the famine phase, where the PHA consumption was made possible due to oxygen production by algae. The internal cycling of carbohydrates was also observed, which was likely linked to bacterial glycogen being used as an additional source of energy for acetate uptake during the feast phase, and restored in the famine phase via PHA degradation. The MPC reached a PHA content of 20%, with a PHA storage yield per acetate similar to aerobic systems, opening up the possibility of a new sunlight-driven PHA production process without the need for aeration.
Sigma-Aldrich
Nile Blue A, certified by the Biological Stain Commission
Sigma-Aldrich
Nile Blue A, Dye content ≥75 %
Sigma-Aldrich
Nile Blue A perchlorate, Dye content 95 %
Sigma-Aldrich
Nile Blue chloride, Dye content 85 %