Merck
  • Investigation of the interactions of lupane type pentacyclic triterpenes with outer leaflet membrane phospholipids--Langmuir monolayer and synchrotron X-ray scattering study.
Investigation of the interactions of lupane type pentacyclic triterpenes with outer leaflet membrane phospholipids--Langmuir monolayer and synchrotron X-ray scattering study.
Journal of colloid and interface science (2012-06-12)
Marcin Broniatowski, Michał Flasiński, Paweł Wydro
ABSTRACT
Lupane type pentacyclic triterpenes (LTs) are pharmacologically active natural products isolated from different plants. They have broad spectrum of therapeutic action ranging from anticancer via anti-HIV, antibiotic to anti-inflammatory and anti-protozoal activity. Many scientific papers underline that the key stage in the LT mechanism of action is their incorporation into cellular membrane and the interaction with the structural lipids. In our research we apply Langmuir monolayers as a versatile platform for the investigation of these phenomena, since till now important aspects concerning this issue are incomprehensible. We focus our attention on the interactions of lupeol and betulinic acid with choline-headgroup structural lipids: a representative of saturated glycerophosphatidylcholines (DPPCs), and octadecyl-sphingomyelin--a representative of membrane sphingolipids. Application of complementary physicochemical techniques such as the Langmuir technique, Brewster angle microscopy, and grazing incidence X-ray diffraction supported by thermodynamic analysis enabled us to investigate the intermolecular interactions in such binary model systems. Our results corroborate that LT is miscible with the outer leaflet membrane phospholipids, both DPPC and SM in the whole range of mole ratios. Moreover, the introduction of LT into the phospholipid film, even in small proportion, leads to the loss of periodical ordering of the phospholipid molecules and the disappearance of the diffraction signal as observed by GIXD. Our results also proved that LT does not form any surface complexes of fixed stoichiometry resembling the well characterized lipid rafts.
Sigma-Aldrich
Lupeol, ≥94%
Supelco
Lupeol, analytical standard