Merck
  • Environmental enrichment prevents the late effect of acute stress-induced fear extinction deficit: the role of hippocampal AMPA-GluA1 phosphorylation.
Environmental enrichment prevents the late effect of acute stress-induced fear extinction deficit: the role of hippocampal AMPA-GluA1 phosphorylation.
Translational psychiatry (2021-01-09)
Leonardo Santana Novaes, Letícia Morais Bueno-de-Camargo, Carolina Demarchi Munhoz
ABSTRACT
The persistence of anxiety and the deficit of fear memory extinction are both phenomena related to the symptoms of a trauma-related disorder, such as post-traumatic stress disorder (PTSD). Recently we have shown that single acute restraint stress (2 h) in rats induces a late anxiety-related behavior (observed ten days after stress), whereas, in the present work, we found that the same stress impaired fear extinction in animals conditioned ten days after stress. Fourteen days of environmental enrichment (EE) prevented the deleterious effect of stress on fear memory extinction. Additionally, we observed that EE prevented the stress-induced increase in AMPA receptor GluA1 subunit phosphorylation in the hippocampus, but not in the basolateral amygdala complex and the frontal cortex, indicating a potential mechanism by which it exerts its protective effect against the stress-induced behavioral outcome.
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NMDAR 2B (984-1104) Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 845, Chemicon®, from rabbit