Merck
  • WITHDRAWN: The chromatin remodeling complex NURF localizes to gene bodies and is required for mRNA processing.
WITHDRAWN: The chromatin remodeling complex NURF localizes to gene bodies and is required for mRNA processing.
The Journal of biological chemistry (2018-08-25)
Aiman S Alhazmi, Marissa Mack, Alexander Petencin, Humberto Nelson, Tiffany Rolle, Jordan Hiegel, Syed Haqqani, Nga Dao, Farheen Zaman, Nak-Kyeong Kim, Neel Scarsdale, Charles Lyons, Joseph W Landry
ABSTRACT
This article has been withdrawn by Aiman Alhazmi, Marissa Mack, Tiffany Rolle, Jordan Hiegel, Syed Haqqani, Nga Dao, Farheen Zaman, Nak-Kyeong Kim, Neel Scarsdale, Charles Lyons, and Joseph Landry. Some of the genome-wide data sets were flawed and were not analyzed correctly. The withdrawing authors are in the process of correcting the data sets and re-analyzing them for resubmission.
Sigma-Aldrich
Anti-BPTF (VCU19), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-BPTF Antibody, from rabbit, purified by affinity chromatography