
Testosterone propionate for system suitability

Synonym(s):
Testosterone propionate, 17β-Hydroxy-4-androsten-3-one 17-propionate, 17β-Propionyloxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one 17-propionate
Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing