
Sodium pyruvate-13C3

Synonym(s):
Pyruvic acid-13C3 sodium salt
Linear Formula:
13CH313CO13CO2Na
CAS Number:
Molecular Weight:
113.02
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing