
Piretanide for system suitability

Synonym(s):
Piretanide, 4-Phenoxy-3-(1-pyrrolidinyl)-5-sulfamoylbenzoic acid
Empirical Formula (Hill Notation):
C17H18N2O5S
CAS Number:
Molecular Weight:
362.40
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing