
Glucobrassicin potassium salt

Synonym(s):
3-Indolylmethyl glucosinolate potassium salt
Empirical Formula (Hill Notation):
C16H19KN2O9S2
CAS Number:
Molecular Weight:
486.56
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing