
Fluoxymesterone CIII

Synonym(s):
Fluoxymesterone, 11β,17β-Dihydroxy-9α-fluoro-17α-methyl-4-androsten-3-one, 9α-Fluoro-11β,17β-dihydroxy-17α-methylandrost-4-en-3-one, 9α-Fluoro-11β-hydroxy-17α-methyltestosterone
Empirical Formula (Hill Notation):
C20H29FO3
CAS Number:
Molecular Weight:
336.44
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing