
D-Luciferin sodium salt

Synonym(s):
(S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid sodium salt, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt, Firefly luciferin sodium salt
Empirical Formula (Hill Notation):
C11H7N2NaO3S2
CAS Number:
Molecular Weight:
302.30
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing