Skip to Content
Merck
Divinyl sulfone contains hydroquinone as inhibitor, ≥96%

Divinyl sulfone

Divinyl sulfone contains hydroquinone as inhibitor, ≥96%
Synonym(s):
Vinyl sulfone, Divinyl sulfone
Linear Formula:
(CH2=CH)2SO2
CAS Number:
Molecular Weight:
118.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2