
Dihydromethysticin

Synonym(s):
(+)-Dihydromethysticin, (6S)-6-[2-(1,3-Benzodioxol-5-yl)ethyl]-5,6-dihydro-4-methoxy-2H-pyran-2-one, Dihydromethysticin
Empirical Formula (Hill Notation):
C15H16O5
CAS Number:
Molecular Weight:
276.28
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing