
Alanine

Synonym(s):
L-Alanine, (S)-2-Aminopropionic acid, L-α-Aminopropionic acid
Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing