
1-(4-Chlorophenyl)-1-phenylethanol

Empirical Formula (Hill Notation):
C14H13ClO
CAS Number:
Molecular Weight:
232.71
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing