
12-Deoxywithastramonolide

Synonym(s):
(5α,6α,7α,22R)-6,7-Epoxy-5,22,27-trihydroxy-1-oxoergosta-2,24-dien-26-oic acid δ-lactone, 27-Hydroxywithanolide B, Baimantuoluoside C aglycone
Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing