AP10045S0
Canisters
pkg of (10 Canisters of 100 absorbent pads), sterile, diam. 45 mm
About This Item
material
cellulose pad(s)
clear device
polystyrene
sterility
sterile
packaging
pkg of (10 Canisters of 100 absorbent pads)
diam.
45 mm
input
sample(s) (environmental)
sample type sport drink
sample type beverage(s)
sample type food(s)
sample type bottled water
sample type cider
sample type cosmetics
sample type soft drink
sample type beer
sample type wine
application(s)
bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical
shipped in
ambient
General description
Application
Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method.
Linkage
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service