Skip to Content
Merck
All Photos(1)

Key Documents

1.11799

Supelco

Sulfanilamide

GR for analysis Reag. Ph Eur

Synonym(s):

Sulfanilamide, 4-Aminobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2S
CAS Number:
Molecular Weight:
172.20
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-563-4
NACRES:
NB.21

Agency

reag. Ph. Eur.

Quality Level

vapor pressure

0.00001 hPa ( 70 °C)

form

solid

potency

3900 mg/kg LD50, oral (Rat)

pH

5.8-6.1 (20 °C, 5 g/L in H2O)

mp

162.8 °C

density

1.54 g/cm3 at 20.3 °C

bulk density

500 kg/m3

storage temp.

15-25°C

InChI

1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

FDDDEECHVMSUSB-UHFFFAOYSA-N

Linkage

Replaces: SX1150-2; SX1150

Analysis Note

Assay (bromometric, calc. on dried substance): ≥ 99
Identity (IR-spectrum): passes test
Melting range (lower value): 163 - 166
Melting range (upper value): 163 - 166
Alkali-insoluble impurities: passes test
Acid-insoluble impurities: passes test
Chloride (Cl): ≤ 0.01 %
Sulphate (SO₄): ≤ 0.02 %
Heavy metals (as Pb): ≤ 0.002 %
Sulfated ash: ≤ 0.1 %
Loss on Drying (105°C): ≤ 0.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service