Merck
  • Systematic evaluation of the conformational properties of aliphatic omega-methoxy methyl esters.
Systematic evaluation of the conformational properties of aliphatic omega-methoxy methyl esters.
The Journal of organic chemistry (2005-09-10)
Carlos Alemán, Jordi Casanovas, H K Hall
ABSTRACT
[structure: see text] A systematic conformational study of omega-methoxy methyl esters, CH(3)O-(CH2)n-COO-CH3 with n = 3 and 4, has been performed using quantum mechanical calculations at the MP2 level. Calculations have been carried out in both gas phase and chloroform solution, a polarizable continuum solvation model being used to represent the latter. Results have been compared with those recently obtained for the analogues omega-hydroxy acids, HO-(CH2)n-COOH with n = 3 and 4. The compounds with n = 3 clearly favor coiled conformations, the population expected for extended and semiextended conformations being very low. However, for compounds with n = 4 the minimum energy extended and semiextended structures become considerably more stable. The overall results indicate that the conformational preferences of the central aliphatic segment of omega-methoxy methyl esters and omega-hydroxy acids are not influenced by the formation of intramolecular O-H...O hydrogen bonds.
Sigma-Aldrich
Methyl 4-methoxybutyrate, 98%
Pricing and availability is not currently available.