Merck
Flucytosine (5-fluorocytosine).
Clinics in laboratory medicine (1987-09-01)
B Gerson
ABSTRACT
Monitoring of flucytosine (5-FC) concentration in serum is of great importance. It may be anticipated that demand for this service will increase in the near future, because this drug is used in situations involving immunocompromised patients, including those afflicted with AIDS.
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog
Flucytosine, European Pharmacopoeia (EP) Reference Standard