Merck
  • The deiodinase family: selenoenzymes regulating thyroid hormone availability and action.
The deiodinase family: selenoenzymes regulating thyroid hormone availability and action.
Cellular and molecular life sciences : CMLS (2001-02-24)
J Köhrle
ABSTRACT
Thyroid hormones control growth, development, differentiation and metabolism in vertebrates. Most of the actions of the active thyroid hormone T3 (3,5,3'-triiodo-L-thyronine) are exerted via ligand-activated nuclear T3 receptors. Activation of the secretory product of the thyroid gland, L-thyroxine (3,3',5,5'-tetraiodo-L-thyronine), or T4, is catalyzed by two enzymes, iodothyronine-5'-deiodinases type I and type II. Inactivation of T4 and T3 occurs via type III iodothyronine-5-deiodinase and to some extent by type I 5'deiodinase. Complementary DNAs (cDNAs) encoding the substrate-binding selenocysteine-containing subunits of the deiodinases were cloned, though some controversy still exists on the type II 5'-deiodinase subunits. Characterization of tissue-specific expression patterns indicates that these selenium-dependent enzymes exert tight control on local and systemic availability of active T3. Thus, deiodinases are envisaged as guardians to the gate of thyroid hormone action mediated by T3 receptors.