Merck
  • A novel biotransformation of 2-formyl-6-naphthoic acid to 2,6-naphthalene dicarboxylic acid by Pseudomonas sp. for the purification of crude 2,6-naphthalene dicarboxylic acid.
A novel biotransformation of 2-formyl-6-naphthoic acid to 2,6-naphthalene dicarboxylic acid by Pseudomonas sp. for the purification of crude 2,6-naphthalene dicarboxylic acid.
Biotechnology letters (2007-10-05)
Dong-Sung Kim, Sung-Kyoon Kim, Yong-Bok Choi, Ik-Hyeon Kwon, Kwan-Hwa Park
ABSTRACT
Crude 2,6-naphthalene dicarboxylic acid was purified by Pseudomonas sp. HN-72 which biotransformed the major impurity of 2-formyl-6-naphthoic acid into 2,6-naphthalene dicarboxylic acid. The biotransformation yield reached 100% when the reaction was performed at 40 degrees C for 1 h, in 200 ml KH(2)PO(4)/KOH buffer (50 mM, pH 8.0), with 0.2% (w/v) crude 2,6-naphthalene dicarboxylic acid and 2.5 mg dry cell wt/ml.
Sigma-Aldrich
2,6-Naphthalenedicarboxylic acid, 99%
Sigma-Aldrich
2,6-Naphthalenedicarboxylic acid, 95%