Merck
  • Synthesis, biological evaluation, and molecular modeling investigation of chiral phenoxyacetic acid analogues with PPARalpha and PPARgamma agonist activity.
Synthesis, biological evaluation, and molecular modeling investigation of chiral phenoxyacetic acid analogues with PPARalpha and PPARgamma agonist activity.
ChemMedChem (2007-03-16)
Giuseppe Fracchiolla, Antonio Laghezza, Luca Piemontese, Giuseppe Carbonara, Antonio Lavecchia, Paolo Tortorella, Maurizio Crestani, Ettore Novellino, Fulvio Loiodice
ABSTRACT
Peroxisome proliferator-activated receptors (PPARs) are ligand-activated transcription factors that govern lipid and glucose homeostasis, and play a central role in cardiovascular disease, obesity, and diabetes. Thus, there is significant interest in developing new and specific agonists for these receptors. Herein we present screening results for a series of chiral phenoxyacetic acid analogues, some of which are potent PPARalpha agonists as well as PPARgamma agonists. The stereochemistry of these compounds plays an important role in determining their activity; the S isomers were observed to be more active than the corresponding R isomers. Interestingly, for one of these analogues, the stereoselectivity toward PPARalpha was reversed, and for this reason docking experiments were performed to rationalize this peculiar behavior.
Product Description
Supelco
Phenoxyacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Phenoxyacetic acid, 98%