Merck
  • Sleep-dependent engram reactivation during hippocampal memory consolidation associated with subregion-specific biosynthetic changes.
Sleep-dependent engram reactivation during hippocampal memory consolidation associated with subregion-specific biosynthetic changes.
iScience (2024-03-25)
Lijing Wang, Lauren Park, Weisheng Wu, Dana King, Alexis Vega-Medina, Frank Raven, Jessy Martinez, Amy Ensing, Katherine McDonald, Zhongying Yang, Sha Jiang, Sara J Aton
ABSTRACT
Post-learning sleep is essential for hippocampal memory processing, including contextual fear memory consolidation. We labeled context-encoding engram neurons in the hippocampal dentate gyrus (DG) and assessed reactivation of these neurons after fear learning. Post-learning sleep deprivation (SD) selectively disrupted reactivation of inferior blade DG engram neurons, linked to SD-induced suppression of neuronal activity in the inferior, but not superior DG blade. Subregion-specific spatial profiling of transcripts revealed that transcriptomic responses to SD differed greatly between hippocampal CA1, CA3, and DG inferior blade, superior blade, and hilus. Activity-driven transcripts, and those associated with cytoskeletal remodeling, were selectively suppressed in the inferior blade. Critically, learning-driven transcriptomic changes differed dramatically between the DG blades and were absent from all other regions. Together, these data suggest that the DG is critical for sleep-dependent memory consolidation, and that the effects of sleep loss on the hippocampus are highly subregion-specific.
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), highly cross-adsorbed, CF555 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 633 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody