Merck
Macrophage Immune Memory Controls Endometriosis in Mice and Humans.
Cell reports (2020-11-05)
Mohamed Jeljeli, Luiza G C Riccio, Sandrine Chouzenoux, Fabiana Moresi, Laurie Toullec, Ludivine Doridot, Carole Nicco, Mathilde Bourdon, Louis Marcellin, Pietro Santulli, Mauricio S Abrão, Charles Chapron, Frédéric Batteux
ABSTRACT
Endometriosis is a frequent, chronic, inflammatory gynecological disease characterized by the presence of ectopic endometrial tissue causing pain and infertility. Macrophages have a central role in lesion establishment and maintenance by driving chronic inflammation and tissue remodeling. Macrophages can be reprogrammed to acquire memory-like characteristics after antigenic challenge to reinforce or inhibit a subsequent immune response, a phenomenon termed "trained immunity." Here, whereas bacille Calmette-Guérin (BCG) training enhances the lesion growth in a mice model of endometriosis, tolerization with repeated low doses of lipopolysaccharide (LPSlow) or adoptive transfer of LPSlow-tolerized macrophages elicits a suppressor effect. LPSlow-tolerized human macrophages mitigate the fibro-inflammatory phenotype of endometriotic cells in an interleukin-10 (IL-10)-dependent manner. A history of severe Gram-negative infection is associated with reduced infertility duration and alleviated symptoms, in contrast to patients with Gram-positive infection history. Thus, the manipulation of innate immune memory may be effective in dampening hyper-inflammatory conditions, opening the way to promising therapeutic approaches.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-STAT3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution