Merck
  • A Novel Sirtuin-3 Inhibitor, LC-0296, Inhibits Cell Survival and Proliferation, and Promotes Apoptosis of Head and Neck Cancer Cells.
A Novel Sirtuin-3 Inhibitor, LC-0296, Inhibits Cell Survival and Proliferation, and Promotes Apoptosis of Head and Neck Cancer Cells.
Anticancer research (2016-01-02)
Turki Y Alhazzazi, Pachiyappan Kamarajan, Yanli Xu, Teng Ai, Liqiang Chen, Eric Verdin, Yvonne L Kapila
ABSTRACT
The survival rate of patients with head and neck squamous cell carcinoma (HNSCC) stands at approximately 50% and this has not improved in decades. This study developed a novel sirtuin-3 (SIRT3) inhibitor (LC-0296) and examined its role in altering HNSCC tumorigenesis. The effect of the SIRT3 inhibitor, LC-0296, on cell survival, proliferation, and apoptosis, and reactive oxygen species levels in HNSCC cells were studied. LC-0296 reduces cell proliferation and promotes apoptosis of HNSCC cells but not of normal human oral keratinocytes. This inhibitory effect is mediated, in part, via modulation of reactive oxygen species levels. Additionally, LC-0296 works synergistically to increase the sensitivity of HNSCC cells to radiation and cisplatin treatment. Development of novel SIRT3 inhibitors, such as LC-0296, might enable the development of new targeted therapies to treat and improve the survival rate of patients with head and neck cancer.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Mitochondria/Cytosol Fractionation Kit, This Mitochondria/Cytosol Fractionation Kit provides reagents for quick & efficient isolation of intact mitochondria from cultured cells.
Pricing and availability is not currently available.