Merck
  • Exploiting Ancillary Ligation To Enable Nickel-Catalyzed C-O Cross-Couplings of Aryl Electrophiles with Aliphatic Alcohols.
Exploiting Ancillary Ligation To Enable Nickel-Catalyzed C-O Cross-Couplings of Aryl Electrophiles with Aliphatic Alcohols.
Journal of the American Chemical Society (2018-03-31)
Preston M MacQueen, Joseph P Tassone, Carlos Diaz, Mark Stradiotto
ABSTRACT
The use of (L)Ni( o-tolyl)Cl precatalysts (L = PAd-DalPhos or CyPAd-DalPhos) enables the C( sp2)-O cross-coupling of primary, secondary, or tertiary aliphatic alcohols with (hetero)aryl electrophiles, including unprecedented examples of such nickel-catalyzed transformations employing (hetero)aryl chlorides, sulfonates, and pivalates. In addition to offering a competitive alternative to palladium catalysis, this work establishes the feasibility of utilizing ancillary ligation as a complementary means of promoting challenging nickel-catalyzed C( sp2)-O cross-couplings, without recourse to precious-metal photoredox catalytic methods.
Sigma-Aldrich
(CyPAd-DalPhos)NiCl(otol), ≥95%