
Tianeptine impurity A

Synonym(s):
Ethyl 7-bromoheptanoate
Linear Formula:
Br(CH2)6CO2C2H5
CAS Number:
Molecular Weight:
237.13
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing