
L-Cysteine S-sulfate sodium salt sesquihydrate

Synonym(s):
L-Cysteine S-sulfate sodium salt sesquihydrate, S-Sulfocysteine, SCys, Sulfo-Cysteine
Empirical Formula (Hill Notation):
C3H6NNaO5S2 · 1.5H2O
CAS Number:
Molecular Weight:
250.23
Products
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
Optical Isomer
Reference Material Type