
Aprepitant for system suitability

Synonym(s):
Aprepitant, 3-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one, 5-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one
Empirical Formula (Hill Notation):
C23H21F7N4O3
CAS Number:
Molecular Weight:
534.43
Products
Formula Weight
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing