Skip to Content
Merck
All Photos(1)

Key Documents

97583

Supelco

N-(2-Aminoethyl)ethanolamine

analytical standard

Synonym(s):

N-(2-Hydroxyethyl)ethylenediamine, 2-(2-Aminoethylamino)ethanol, N-(2-Aminoethyl)ethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NHCH2CH2OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
506012
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

grade

analytical standard

Quality Level

vapor density

3.6 (vs air)

Assay

≥98.0% (GC)

technique(s)

HPLC: suitable

refractive index

n20/D 1.485 (lit.)

bp

238-240 °C/752 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

NCCNCCO

InChI

1S/C4H12N2O/c5-1-2-6-3-4-7/h6-7H,1-5H2

InChI key

LHIJANUOQQMGNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

291.2 °F

Flash Point(C)

144 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service