Merck
HomeProtein ExpressionSomatostatin Receptors
Somatostatin Receptors
Somatostatin (somatotropin release-inhibiting factor, SRIF) is a cyclic peptide widely expressed throughout the CNS, endocrine tissues, and gastrointestinal tract. The 14 amino acid peptide, somatostatin (SRIF-14), was first identified and isolated from ovine hypothalamic extracts as a potent inhibitor of growth hormone release from the pituitary. Subsequently, a longer N-terminally extended form (SRIF-28) was identified. SRIF exerts a wide range of biological actions, including inhibition of secretion of growth hormone, insulin, glucagon, and gastrin as well as other hormones secreted by the pituitary and gastrointestinal tract. Somatostatin also acts as a neuromodulator in the CNS and shows an anti-proliferative effect on a wide range of cancer cells. A rat neuropeptide, named cortistatin (CST), with strong homology to SRIF has also been cloned. Depending on the species, CST is a tetradecapeptide, sharing up to 11 amino acids with SRIF. The human homolog of CST appears to be a heptadecapeptide (CST-17). By analogy with SRIF, there may also be a larger isoform of CST, i.e. CST-29. Although various physiological parameters, including transitions between sleep phases, consolidation of short- and long-term memory and locomotor activity, respond in an apparently transmitter-specific manner to SRIF and CST, the latter is an endogenous ligand at SRIF receptors, with nanomolar affinity for each subtype. The existence of specific CST receptors has not been demonstrated so far, although it was suggested that MGRX2 may play such a role.
SRIF and CST act via a family of G protein-coupled receptors, of which five subtypes (sst1-sst5) have been cloned and characterized from various species. Sequence homology is 39-57% among the five subtypes, each being highly conserved across species. Based on structural and operational features, they can be divided into two groups: SRIF-1 includes sst2, sst3, and sst5 receptors while SRIF-2 consists of sst1 and sst4 receptors, distinguished by the high affinity of cyclic peptides such as octreotide (SMS-201-995), seglitide (MK-678) and lanreotide (BIM-23014) for the SRIF-1 group.
Molecular biological techniques and subtype-selective antibodies have been used to determine the distribution of the sst1-sst5 gene transcripts and receptor protein in brain and periphery. Selective peptidic agonists at sst1 (CH-275) and sst4 receptors (NNC 26-9100) have been described, as has an sst3 receptor antagonist (sst3-ODN-8). However, the use of peptides for in vivo experiments is limited by pharmacokinetic constraints that may be addressed by the recent description of non-peptide agonists selective for each of the five somatostatin receptors. Further, non-peptide antagonists such as SRA880 (sst1 selective), ACQ090 (sst3 selective), and sst4 selective β peptide agonists which all show adequate stability and bioavailability have been described more recently.
Studies utilizing subtype-selective SRIF analogs in both in vivo and in vitro experiments demonstrate that sst2 receptors are the major player in the SRIF receptor family with broad inhibitory effects on the endocrine secretion, e.g. growth hormone, insulin, glucagon, gastrin, cholecystokinin, vasoactive intestinal peptide and secretin, as well as the exocrine secretion, e.g. gastric acid, intestinal fluid, and pancreatic enzymes. Sst2 receptors also seem to play a major role in various forms of gastro-enteropancreatic cancers, in epilepsy and pain. The sst1 receptor may function as an autoreceptor at least in basal ganglia and the eye. Sst3 receptors are enigmatically localized to neuronal cilia, and sst3 antagonists have marked behavioral effects. Sst4 receptors are highly expressed in the lung, but their role remains to be defined, although in the mouse they modulate epileptic activity. Sst5 receptors mediate inhibition of insulin release from the pancreatic β-cells in addition to regulating growth hormone release.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Abbreviations
BIM-23027: c[N-methyl-Ala-Tyr-D-Trp-Lys-Abu-Phe]
BIM-23056: D-Phe-Phe-Tyr-D-Trp-Lys-Val-Phe-D-Nal-NH2
CH-275: des-AA1,2,5(D-Trp8,Iamp9)-SRIF
Cyanamid-154806: AC-4-NO2-Phe-c[D-Cys-Tyr-D-Trp-Lys-Thr-Cys]-D-Tyr-NH2
GEP: Gastro-enteropancreatic
GI: Gastrointestinal
MK-678 (Seglitide): c[N-Methyl-Ala-Tyr-D-Trp-Lys-Val-Phe]
L-797,591: (2R)-N-(6-Amino-2,2,4-trimethylhexyl)-3-(1-naphthyl)-2-({[(2-phenylethyl)(2-pyridin-2-ylethyl)amino]carbonyl}amino)propanamide
L-054,522: tert-Butyl (bS)-b-methyl-N-{[4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]carbonyl}-D-tryptophyl-L-lysinate
L-779,976: (bS)-N-{[3-(Aminomethyl)cyclohexyl]methyl}-b-methyl-N-{[4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]carbonyl}-D-tryptophamide
L-796,778: Methyl (2S)-6-amino-2-[((2R)-2-{[({(1S)-1-benzyl-2-[(4-nitrophenyl)amino]-2-oxoethyl}amino)carbonyl]amino}hexanoyl)amino]hexanoate
L-803,087: Methyl (2S)-5-{[amino(imino)methyl]amino}-2-{[4-(5,7-difluoro-2-phenyl-1H-indol-3-yl)butanoyl]amino}pentanoate or methyl N2-[4-(5,7-difluoro-2-phenyl-1H-indol-3-yl)butanoyl]-L-argininate
L-817,818: (2R)-2-Aminopropyl N2-{[2-(2-naphthyl)-1H-benzo[g]indol-3-yl]acetyl}-L-lysinat
NNC 26-9100: I-[3-[N-(5-Bromopyridin-2-yl)-N-(3,4-dichlorobenzyl)aminopropyl]-3-[3-(1H-imidazol-1-yl)propyl]-thiourea
NVP-ACQ090: [4-(3,4-Difluoro-phenyl)-piperazin-1-yl]-{(4S,4aS,8aR)-2-[(S)-3-(6-methoxy-pyridin-3-yl)-2-methyl-propyl]-decahydro-isoquinolin-4-yl}-methanone
NVP-SRA880: [3R,4aR,10aR]-1,2,3,4,4a,5,10,10a-Octahydro-6-methoxy-1-methyl-benz[g] quinoline-3-carboxylic-acid-4-(4-nitro-phenyl)-piperazine-amide, hydrogen malonate
sst3-ODN-8: Carbamoyl-des-AA1,2,4,5,12,13[D-Cys3,D-Arg18(Me,2-naphthoyl)]-SRIF
[125I]-[Tyr25]-SRIF28: 3-[125I]iodotyrosyl25-SRIF-(Leu8,D-Trp22,Tyr25)
Similar Products
Loading
References
1.
Ankersen M, Crider M, Liu S, Ho B, Andersen HS, Stidsen C. 1998. Discovery of a Novel Non-Peptide Somatostatin Agonist with SST4Selectivity. J. Am. Chem. Soc.. 120(7):1368-1373. https://doi.org/10.1021/ja973325x
2.
Cakir M, Dworakowska D, Grossman A. 2010. Somatostatin receptor biology in neuroendocrine and pituitary tumours: part 1 - molecular pathways. 14(11):2570-2584. https://doi.org/10.1111/j.1582-4934.2010.01125.x
3.
Csaba Z, Peineau S, Dournaud P. 2012. Molecular mechanisms of somatostatin receptor trafficking. 48(1):R1-R12. https://doi.org/10.1530/jme-11-0121
4.
Hasskarl J, Kaufmann M, Schmid HA. 2011. Somatostatin receptors in non-neuroendocrine malignancies: the potential role of somatostatin analogs in solid tumors. Future Oncology. 7(7):895-913. https://doi.org/10.2217/fon.11.66
5.
Hocart SJ, Jain R, Murphy WA, Taylor JE, Coy DH. 1999. Highly Potent Cyclic Disulfide Antagonists of Somatostatin. J. Med. Chem.. 42(11):1863-1871. https://doi.org/10.1021/jm9806289
6.
D H, al. e. 2000. Somatostatin receptors., in: The IUPHAR Compendium of Receptor Characterization and Classification. 2nd edition, pp. 354-364. London, UK: IUPHAR Media.
7.
Hoyer D, Nunn C, Hannon J, Schoeffter P, Feuerbach D, Schuepbach E, Langenegger D, Bouhelal R, Hurth K, Neumann P, et al. 2004. SRA880, in vitro characterization of the first non-peptide somatostatin sst1 receptor antagonist. Neuroscience Letters. 361(1-3):132-135. https://doi.org/10.1016/j.neulet.2004.02.017
8.
Møller LN, Stidsen CE, Hartmann B, Holst JJ. 2003. Somatostatin receptors. Biochimica et Biophysica Acta (BBA) - Biomembranes. 1616(1):1-84. https://doi.org/10.1016/s0005-2736(03)00235-9
9.
Nunn C, Rueping M, Langenegger D, Schuepbach E, Kimmerlin T, Micuch P, Hurth K, Seebach D, Hoyer D. 2003. ?2/?3-di- and ?/?3-tetrapeptide derivatives as potent agonists at somatostatin sst4 receptors. Naunyn-Schmiedeberg's Arch Pharmacol. 367(2):95-103. https://doi.org/10.1007/s00210-002-0673-4
10.
Olias G, Viollet C, Kusserow H, Epelbaum J, Meyerhof W. 2004. Regulation and function of somatostatin receptors. J Neurochem. 89(5):1057-1091. https://doi.org/10.1111/j.1471-4159.2004.02402.x
11.
Reubi JC, Schaer J, Wenger S, Hoeger C, Erchegyi J, Waser B, Rivier J. 2000. SST3-selective potent peptidic somatostatin receptor antagonists. Proceedings of the National Academy of Sciences. 97(25):13973-13978. https://doi.org/10.1073/pnas.250483897
12.
Rohrer SP. 1998. Rapid Identification of Subtype-Selective Agonists of the Somatostatin Receptor Through Combinatorial Chemistry. 282(5389):737-740. https://doi.org/10.1126/science.282.5389.737
13.
Siehler S, Seuwen K, Hoyer D. 1998. [125I]Tyr10-cortistatin14 labels all five somatostatin receptors. Naunyn-Schmiedeberg's Arch Pharmacol. 357(5):483-489. https://doi.org/10.1007/pl00005197
14.
Sun L, H. Coy D. 2011. Somatostatin Receptor-Targeted Anti-Cancer Therapy. CDD. 8(1):2-10. https://doi.org/10.2174/156720111793663633
15.
Weckbecker G, Lewis I, Albert R, Schmid HA, Hoyer D, Bruns C. 2003. Opportunities in somatostatin research: biological, chemical and therapeutic aspects. Nat Rev Drug Discov. 2(12):999-1017. https://doi.org/10.1038/nrd1255
16.
Yang L. 1999. Chapter 21. Non-peptide Somatostatin Receptor Ligands.209-218. https://doi.org/10.1016/s0065-7743(08)60583-7