Merck
UHPLC Analysis of Fat Soluble Vitamins on Titan C18, Methanol Mobile Phase
UHPLC Analysis of Fat Soluble Vitamins on Titan C18, Methanol Mobile Phase application for UHPLC
Materials
standard
Product No.
Description
Pricing
DL-α-Tocopherol acetate
≥96% (HPLC)
Retinyl acetate
analytical standard
(±)-α-Tocopherol
analytical standard
(+)-γ-Tocopherol
analytical standard
δ-Tocopherol
analytical standard
Retinyl Palmitate (Vitamin A Palmitate)
Pharmaceutical Secondary Standard; Certified Reference Material
CONDITIONS
column
Titan C18, 50 mm x 2.1 mm I.D., 1.9 μm particles (577122-U)
mobile phase
100% methanol
flow rate
0.4 mL/min
pressure
2741 psi (189 bar)
column temp.
30 °C
injection
UV, 280 nm
injection
0.5 μL
sample
All compounds at 0.5 mg/mL except retinyl acetate at 0.05 mg/mL in methanol
Description
Legal Information
null