HPLC Analysis of Diet Soda Additives on Ascentis® Express RP-Amide
![HPLC Analysis of Diet Soda Additives on Ascentis® Express RP-Amide application for HPLC](/deepweb/assets/sigmaaldrich/product/images/329/685/4a31e3e9-8705-42cd-ac7d-b09f8a4e6671/800/4a31e3e9-8705-42cd-ac7d-b09f8a4e6671.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express RP-Amide, 3 cm x 4.6 mm I.D., 2.7 μm particles (53921-U)
mobile phase
[A] 20 mM ammonium acetate, pH 5.6 with acetic acid; [B] water; [C] acetonitrile
gradient
20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
pressure
3336 psi (230 bar)
flow rate
3.0 mL/min
column temp.
40 °C
detector
UV, 214 nm
injection
1 μL
sample
Diet Soda 100 - 500 μg/mL in buffer
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany