HPLC Analysis of Chlorthalidone Enantiomers on Astec® CYCLOBOND I 2000 RSP
![HPLC Analysis of Chlorthalidone Enantiomers on Astec® CYCLOBOND I 2000 RSP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/280/999/de31e79e-7790-4921-9ee0-ba7ae7bebb88/800/de31e79e-7790-4921-9ee0-ba7ae7bebb88.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000 RSP, 25 cm x 4.6 mm I.D., 5 μm particles (20324AST)
mobile phase
[A] 1% triethylamine acetate, pH 4.1; [B] acetonitrile; (95:5, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null