Merck
  • Cantharidin Inhibits the Growth of Triple-Negative Breast Cancer Cells by Suppressing Autophagy and Inducing Apoptosis in Vitro and in Vivo.
Cantharidin Inhibits the Growth of Triple-Negative Breast Cancer Cells by Suppressing Autophagy and Inducing Apoptosis in Vitro and in Vivo.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-10-20)
Hong-Chang Li, Zhi-Hua Xia, Ya-Feng Chen, Fan Yang, Wen Feng, Han Cai, Yi Mei, Yi-Ming Jiang, Ke Xu, Dian-Xu Feng
ABSTRACT
Cantharidin, a type of terpenoid secreted by the blister beetle Mylabris phalerata (Pallas), has attracted great attention in cancer therapy because of its potential anti-cancer activities. Here, we report the effects on apoptosis and autophagy in human triple-negative breast cancer (TNBC) cell lines after treatment with cantharidin and attempt to elucidate the underlying mechanisms. MDA-MB-231 and MDA-MB-468 cells were treated with cantharidin and cell proliferation was examined using CCK-8 and clone formation assays. The expression of apoptosis- and autophagy-associated proteins was detected by western blotting. Cells were infected with lentivirus carrying the Beclin-1 gene, and MDA-MB-231-beclin1 (MB231-Bec) and MDA-MB-468-beclin-1(MB468-Bec) cells stably expressing Beclin-1 were established. Autophagic vacuoles in cells were observed with LC3 staining using fluorescence microscopy, and apoptotic cells were detected via flow cytometry. Tumor growth was assessed by subcutaneous inoculation of TNBC cells into BALB/c nude mice. Cantharidin inhibited the proliferation of MDA-MB-231 and MDA-MB-468 cells, and induced cell apoptosis. Cantharidin additionally inhibited the conversion of LC3 I to LC3 II and autophagosome formation by suppressing the expression of Beclin-1. Furthermore, overexpression of Beclin-1 in TNBC cells attenuated the cytotoxicity of cantharidin. In vivo, cantharidin inhibited the growth of MDA-MB-231 and MDA-MB-468 xenografts in nude mice by suppressing autophagy and inducing apoptosis, and Beclin-1 overexpression in TNBC cells reduced the efficacy of cantharidin. Cantharidin inhibits autophagy by suppressing Beclin-1 expression and inducing apoptosis of TNBC cells in vitro and in vivo, thereby representing a potential strategy for the treatment of TNBC.