Merck
  • IFN-λ4 potently blocks IFN-α signalling by ISG15 and USP18 in hepatitis C virus infection.
IFN-λ4 potently blocks IFN-α signalling by ISG15 and USP18 in hepatitis C virus infection.
Scientific reports (2017-06-21)
Pil Soo Sung, Seon-Hui Hong, Jae-Hee Chung, Sojeong Kim, Su-Hyung Park, Ho Min Kim, Seung Kew Yoon, Eui-Cheol Shin
ABSTRACT
Genetic polymorphisms in IFNL4 have been shown to predict responses to IFN-α-based therapy in hepatitis C virus (HCV)-infected patients. The IFNL4-ΔG genotype, which encodes functional IFN-λ4 protein, is associated with a poor treatment response. In the present study, we investigated the induction and biological effects of IFN-λ4 in HCV-infected hepatocytes and their association with responsiveness to IFN-α. We also studied the effects of direct-acting antiviral (DAA) treatment on IFN-λ4 expression and IFN-α responsiveness. HCV infection induced IFN-λ4 expression at mRNA and protein levels in primary human hepatocytes (PHHs). In hepatoma cells, IFNL4 gene transfection or recombinant IFN-λ4 protein treatment robustly increased the protein levels of ISG15 and USP18 in an IFNLR1-dependent manner and potently blocked IFN-α signalling. The ISG15/USP18-mediated IFN-α unresponsiveness was demonstrated by transfection of siRNAs targeting ISG15 and/or USP18. This potent IFN-λ4 effect was related to prolonged ISG expression after IFNL4 gene transfection. DAA treatment of HCV-infected PHHs reduced the expression of IFN-λs, including IFN-λ4, and restored IFN-α responsiveness. These results demonstrate that virus-induced IFN-λ4 potently blocks IFN-α signalling by inducing high protein levels of ISG15 and USP18. Moreover, the data clearly demonstrate that DAA therapy restores IFN-α responsiveness in HCV-infected cells.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Interferon Lambda 4, clone 4G1 Antibody, clone 4G1, 1 mg/mL, from mouse